Learn More
Medchemexpress LLC Guanine hydrochloride | 635-39-2 | MFCD00213670 | 98.8% | 187.59 g/mol | C5H6ClN5O | 50mg
SDP
Catalog No. 5000268103
Encompass_Preferred
Guanine hydrochloride is one of the fundamental components of nucleic acids (DNA and RNA) It is a purine derivative composed of a fused pyrimidine-imidazole ring system with conjugated double bonds Guanine hydrochloride has the potential to serve as a large-capacity nitrogen reservoir[1]
Catalog No. 50-002-68103 Supplier Medchemexpress LLC Supplier No. HYY1055A50MG
Guanine hydrochloride is one of the fundamental components of nucleic acids (DNA and RNA) It is a purine derivative composed of a fused pyrimidine-imidazole ring system with conjugated double bonds Guanine hydrochloride has the potential to serve as a large-capacity nitrogen reservoir[1]