Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanine Sulfate Dihydrate 98.0+%, TCI America™
SDP

Catalog No. G01681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G01681G 1 g
G016825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G01681G Supplier TCI America Supplier No. G01681G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10333-92-3
Molecular Formula C5H5N5O
MDL Number MFCD00213671
Synonym 2-Aminohypoxanthine Sulfate

Specifications

Color White
Quantity 1 g
Formula Weight 400.33
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Guanine Sulfate Dihydrate
EINECSNumber (9)-0328
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.