Learn More
Medchemexpress LLC Guanosine-1′-13C, 2′-deoxy-, monohydrate | 478511-26-1 | 99.9% | 286.25 | C913CH15N5O5 | 1 MG
SDP
Catalog No. 5000280936
Encompass_Preferred
2'-Deoxyguanosine-13C monohydrate is a 13C-labeled form of the nucleoside 2'-deoxyguanosine supplied as the monohydrate for analytical and research applications. It is intended for use as a stable-isotope internal standard, tracer, or reference material in mass spectrometry and metabolic studies. Certificate of analysis and safety data sheet are available for lot-specific purity and storage information.
  • Isotope-labeled (13C) monohydrate suitable for analytical standards and tracing.
  • High purity: 99.9% by HPLC, reported on the certificate of analysis.
  • Supplied as a solid in small milligram quantities for laboratory use.
  • Common applications include LC-MS/MS internal standard and metabolic labeling.
  • Certificate of analysis and safety data sheet available for handling and storage instructions.
Catalog No. 50-002-80936 Supplier Medchemexpress LLC Supplier No. HYW338294S21MG
2'-Deoxyguanosine-13C monohydrate is a 13C-labeled form of the nucleoside 2'-deoxyguanosine supplied as the monohydrate for analytical and research applications. It is intended for use as a stable-isotope internal standard, tracer, or reference material in mass spectrometry and metabolic studies. Certificate of analysis and safety data sheet are available for lot-specific purity and storage information.
  • Isotope-labeled (13C) monohydrate suitable for analytical standards and tracing.
  • High purity: 99.9% by HPLC, reported on the certificate of analysis.
  • Supplied as a solid in small milligram quantities for laboratory use.
  • Common applications include LC-MS/MS internal standard and metabolic labeling.
  • Certificate of analysis and safety data sheet available for handling and storage instructions.