Learn More
Medchemexpress LLC Guanosine-15N5 | 99.6% | 288.21 | 1 MG
SDP
Catalog No. 5000427693
Encompass_Preferred
Guanosine-15N5 is an isotopically labeled form of Guanosine, where 15N (nitrogen-15) replaces the natural nitrogen atoms in the molecule. Guanosine is a purine nucleoside, formed by the attachment of guanine to a ribose sugar via a β-N9-glycosidic bond. This compound is primarily intended for research purposes.
  • 15N labeled compound
  • Purine nucleoside
  • Formed from guanine and ribose
  • Possesses anti-HSV activity
  • Suitable for research applications
Catalog No. 50-004-27693 Supplier Medchemexpress LLC Supplier No. HYN0097S31MG
Guanosine-15N5 is an isotopically labeled form of Guanosine, where 15N (nitrogen-15) replaces the natural nitrogen atoms in the molecule. Guanosine is a purine nucleoside, formed by the attachment of guanine to a ribose sugar via a β-N9-glycosidic bond. This compound is primarily intended for research purposes.
  • 15N labeled compound
  • Purine nucleoside
  • Formed from guanine and ribose
  • Possesses anti-HSV activity
  • Suitable for research applications