Learn More
Medchemexpress LLC HY-N0097 1g Medchemexpress, Guanosine CAS:118-00-3 Purity:>98%
SDP
Catalog No. 501933391
Encompass_Preferred
Medchemexpress, HY-N0097 1g Guanosine CAS:118-00-3 Guanosine is a purine nucleoside comprising guanine attached to a ribose (ribofuranose) ring via a β-N9-glycosidic bond. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3391 Supplier Medchemexpress LLC Supplier No. HYN00971G
Medchemexpress, HY-N0097 1g Guanosine CAS:118-00-3 Guanosine is a purine nucleoside comprising guanine attached to a ribose (ribofuranose) ring via a β-N9-glycosidic bond. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.