Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine, 2′-deoxy-N-methyl | 19916-77-9 | 99.8% | 281.27 | C11H15N5O4 | 10 MG
SDP

Catalog No. 5000285970
Encompass_Preferred

2′-Deoxy-N-methylguanosine is a purine nucleoside analog supplied as a solid for research use. It is a 2′-deoxy derivative of N-methylguanosine and is used in studies of modified nucleosides, DNA adducts, enzymatic assays, and nucleic acid chemistry.

  • Purity 99.8% for reliable experimental results.
  • Molecular weight 281.27 and formula C11H15N5O4 for precise molecular characterization.
  • Available in multiple milligram quantities to support small-scale experiments.
  • Solid form for stable storage and accurate weighing.
  • SMILES provided for computational chemistry and database indexing.
  • Suitable for biochemical and nucleic acid research applications.

Catalog No. 50-002-85970 Supplier Medchemexpress LLC Supplier No. HY15440610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2′-Deoxy-N-methylguanosine is a purine nucleoside analog supplied as a solid for research use. It is a 2′-deoxy derivative of N-methylguanosine and is used in studies of modified nucleosides, DNA adducts, enzymatic assays, and nucleic acid chemistry.

  • Purity 99.8% for reliable experimental results.
  • Molecular weight 281.27 and formula C11H15N5O4 for precise molecular characterization.
  • Available in multiple milligram quantities to support small-scale experiments.
  • Solid form for stable storage and accurate weighing.
  • SMILES provided for computational chemistry and database indexing.
  • Suitable for biochemical and nucleic acid research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.