Learn More
Selleck Chemical LLC Guanosine S2439-1g
SDP
Catalog No. 503111741
Encompass_Preferred
Guanosine is a purine nucleoside comprising guanine attached to a ribose (ribofuranose) ring via a -N9-glycosidic bond
Catalog No. 50-311-1741 Supplier Selleck Chemical LLC Supplier No. S24391G
Guanosine is a purine nucleoside comprising guanine attached to a ribose (ribofuranose) ring via a -N9-glycosidic bond