Learn More
TARGETMOL CHEMICALS INC Research Chemical 25mg
SDP
Supplier:  TARGETMOL CHEMICALS INC T506525MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg, 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GUANOSINE 3',5'-CYCLIC MONOPHOSPHATE SOD (cGMP sodium salt) , also known as cGMP, is a cellular regulatory agent and has been described as a second messenger. Its levels increase in response to a variety of hormones, including acetylcholine, insulin, and oxytocin. GUANOSINE 3',5'-CYCLIC MONOPHOSPHATE SOD activates protein kinase G (PKG) and modulates ion channel conductance, with signaling affecting diverse processes including smooth muscle relaxation and proliferation, phototransduction, and energy homeostasis. purity: 99%
Catalog No. 50-303-8149