Learn More
Medchemexpress LLC Guanosine 3',5'-cyclic monophosphate (13C,15N2) | MFCD00070129 | 348.18 g·mol⁻1 | C913CH12N315N2O7P | 500 UG
SDP
Catalog No. 5000283934
Encompass_Preferred
Cyclic GMP-13C,15N2 is a stable-isotope labeled guanosine 3',5'-cyclic monophosphate (cGMP) with 13C and 15N substitutions, supplied as a 500 μg vial. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and metabolic studies of cGMP signaling. Molecular weight 348.18 g·mol⁻1; refer to the certificate of analysis for purity and storage instructions.
  • Stable-isotope labeled (13C, 15N) for accurate quantitation.
  • Suitable as an internal standard for LC-MS/MS and other analytical methods.
  • Supplied as a 500 μg vial for trace-level assays.
  • Certificate of analysis available for lot-specific purity and storage conditions.
  • Supports studies of nitric oxide and natriuretic peptide signaling pathways.
Catalog No. 50-002-83934 Supplier Medchemexpress LLC Supplier No. HYW654000500UG
Cyclic GMP-13C,15N2 is a stable-isotope labeled guanosine 3',5'-cyclic monophosphate (cGMP) with 13C and 15N substitutions, supplied as a 500 μg vial. It is intended for use as an internal standard or tracer in quantitative mass spectrometry and metabolic studies of cGMP signaling. Molecular weight 348.18 g·mol⁻1; refer to the certificate of analysis for purity and storage instructions.
  • Stable-isotope labeled (13C, 15N) for accurate quantitation.
  • Suitable as an internal standard for LC-MS/MS and other analytical methods.
  • Supplied as a 500 μg vial for trace-level assays.
  • Certificate of analysis available for lot-specific purity and storage conditions.
  • Supports studies of nitric oxide and natriuretic peptide signaling pathways.