Learn More
Medchemexpress LLC Guanosine 3',5'-cyclic monophosphate | 7665-99-8 | MFCD00070129 | 99.5% | 345.21 g/mol | C10H12N5O7P | 5 MG
SDP
Catalog No. 5000264800
Encompass_Preferred
Cyclic GMP (cGMP) is supplied as an analytical reference standard of guanosine 3',5'-cyclic monophosphate in solid form. The material is intended for use as a calibration standard, quality control reference, or reagent in analytical and biochemical workflows that require a high-purity cGMP reference.
  • Analytical reference standard for calibration and quality control.
  • High purity suitable for HPLC and other analytical methods.
  • Solid powder form for accurate weighing and solution preparation.
  • Available in small pack sizes for analytical use, including 5 mg.
  • Suitable for biochemical assays and method development involving cGMP.
Catalog No. 50-002-64800 Supplier Medchemexpress LLC Supplier No. HY113469R5MG
Cyclic GMP (cGMP) is supplied as an analytical reference standard of guanosine 3',5'-cyclic monophosphate in solid form. The material is intended for use as a calibration standard, quality control reference, or reagent in analytical and biochemical workflows that require a high-purity cGMP reference.
  • Analytical reference standard for calibration and quality control.
  • High purity suitable for HPLC and other analytical methods.
  • Solid powder form for accurate weighing and solution preparation.
  • Available in small pack sizes for analytical use, including 5 mg.
  • Suitable for biochemical assays and method development involving cGMP.