Learn More
Medchemexpress LLC HY-113066A 10mg Medchemexpress, Guanosine 5'-diphosphate disodium salt CAS:7415-69-2 Purity:>98%
SDP
Catalog No. 501932450
Encompass_Preferred
Medchemexpress, HY-113066A 10mg Guanosine 5'-diphosphate disodium salt CAS:7415-69-2 Guanosine 5'-diphosphate disodium salt is a nucleoside diphosphate. Guanosine 5'-diphosphate is a potential iron mobilizer, which prevents the hepcidin-ferroportin interaction and modulating the interleukin-6 (IL-6)/stat-3 pathway [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2450 Supplier Medchemexpress LLC Supplier No. HY113066A10MG
Medchemexpress, HY-113066A 10mg Guanosine 5'-diphosphate disodium salt CAS:7415-69-2 Guanosine 5'-diphosphate disodium salt is a nucleoside diphosphate. Guanosine 5'-diphosphate is a potential iron mobilizer, which prevents the hepcidin-ferroportin interaction and modulating the interleukin-6 (IL-6)/stat-3 pathway [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.