Learn More
Medchemexpress LLC Guanosine 5'-diphosphate-13C10 dilithium | 98.7% | 461.97 | 1 MG
SDP
Catalog No. 5000435655
Encompass_Preferred
Guanosine 5'-diphosphate-13C10 (GDP-13C10) dilithium is a 13C-labeled nucleoside diphosphate. It activates adenosine 5'-triphosphate-sensitive K+ channels and acts as a potential iron mobilizer, preventing hepcidin-ferroportin interaction and modulating the interleukin-6 (IL-6)/stat-3 pathway. This compound is used in inflammation research, including anemia of inflammation (AI).
  • Activates adenosine 5'-triphosphate-sensitive K+ channels.
  • Acts as a potential iron mobilizer.
  • Prevents hepcidin-ferroportin interaction.
  • Modulates the interleukin-6 (IL-6)/stat-3 pathway.
  • Utilized in inflammation research, including anemia of inflammation (AI).
  • Functions as a tracer for quantitative analysis.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes like 13C for drug development quantitation.
Catalog No. 50-004-35655 Supplier Medchemexpress LLC Supplier No. HY113066S11MG
Guanosine 5'-diphosphate-13C10 (GDP-13C10) dilithium is a 13C-labeled nucleoside diphosphate. It activates adenosine 5'-triphosphate-sensitive K+ channels and acts as a potential iron mobilizer, preventing hepcidin-ferroportin interaction and modulating the interleukin-6 (IL-6)/stat-3 pathway. This compound is used in inflammation research, including anemia of inflammation (AI).
  • Activates adenosine 5'-triphosphate-sensitive K+ channels.
  • Acts as a potential iron mobilizer.
  • Prevents hepcidin-ferroportin interaction.
  • Modulates the interleukin-6 (IL-6)/stat-3 pathway.
  • Utilized in inflammation research, including anemia of inflammation (AI).
  • Functions as a tracer for quantitative analysis.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes like 13C for drug development quantitation.