Learn More
Medchemexpress LLC Guanosine 5'-diphosphate (disodium salt) | 7415-69-2 | MFCD00084665 | 98.2% | 487.16 | C10H13N5Na2O11P2 | 5 MG
SDP
Catalog No. 5000262974
Encompass_Preferred
Guanosine 5'-diphosphate disodium salt is a purine nucleoside diphosphate used in biochemical and cellular research to probe GTPase activity, modulate ATP-sensitive K+ channels, and investigate iron mobilization pathways. Provided as a white to off-white solid with high purity, it is suitable for analytical assays and biochemical studies when stored under recommended conditions.
  • High purity suitable for analytical and biochemical assays.
  • Solid form convenient for storage, weighing, and long-term use.
  • Supports studies of GTPase kinetics and ATP-sensitive K+ channel modulation.
  • Applicable for research on iron mobilization and hepcidin-ferroportin interactions.
  • Stable under recommended storage: -20°C for solids; in solution, -80°C up to 6 months.
Catalog No. 50-002-62974 Supplier Medchemexpress LLC Supplier No. HY113066A5MG
Guanosine 5'-diphosphate disodium salt is a purine nucleoside diphosphate used in biochemical and cellular research to probe GTPase activity, modulate ATP-sensitive K+ channels, and investigate iron mobilization pathways. Provided as a white to off-white solid with high purity, it is suitable for analytical assays and biochemical studies when stored under recommended conditions.
  • High purity suitable for analytical and biochemical assays.
  • Solid form convenient for storage, weighing, and long-term use.
  • Supports studies of GTPase kinetics and ATP-sensitive K+ channel modulation.
  • Applicable for research on iron mobilization and hepcidin-ferroportin interactions.
  • Stable under recommended storage: -20°C for solids; in solution, -80°C up to 6 months.