Learn More
Abcam Guanosine 5'-diphosphate [GDP] disodium salt, Nucleoside diphosphate, 100MG
Catalog No. 502583226
Encompass_Preferred
MW 487.17 g/mol, Purity >98%. Nucleoside diphosphate used as a substrate of pyruvate kinase to produce GTP in support of RNA biosynthesis. Used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors (GPCR). Also used to study cell signaling processes mediate by guanine nucleotide exchange factors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3226 Supplier Abcam Supplier No. AB146529100MG
MW 487.17 g/mol, Purity >98%. Nucleoside diphosphate used as a substrate of pyruvate kinase to produce GTP in support of RNA biosynthesis. Used to study the kinetics and characteristics of GTPases such as those associated with G-protein coupled receptors (GPCR). Also used to study cell signaling processes mediate by guanine nucleotide exchange factors.
The product is subject to the following: Abcam Restricted Use Statement