Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine 5'-diphosphate, monosodium salt | 43139-22-6 | MFCD00070121 | 96.0% | 465.18 g/mol | C10H14N5NaO11P2 | 5 MG
SDP

Catalog No. 5000264190
Encompass_Preferred

Guanosine 5'-diphosphate (sodium) is a nucleoside diphosphate used in biochemical and cellular research. It acts as a ligand for nucleotide-binding proteins, can activate ATP-sensitive K+ channels, and is applied in studies of inflammation and iron mobilization pathways.

  • Monosodium salt form suitable for aqueous and enzymatic studies.
  • Nominal purity 96.0% by HPLC; batch certificate of analysis may report higher purity.
  • White to off-white solid appearance; stable under recommended storage conditions.
  • Used in inflammation research, including hepcidin-ferroportin and IL-6/STAT3 pathway studies.
  • Molecular formula C10H14N5NaO11P2; molecular weight 465.18 g/mol.

Catalog No. 50-002-64190 Supplier Medchemexpress LLC Supplier No. HY113066C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanosine 5'-diphosphate (sodium) is a nucleoside diphosphate used in biochemical and cellular research. It acts as a ligand for nucleotide-binding proteins, can activate ATP-sensitive K+ channels, and is applied in studies of inflammation and iron mobilization pathways.

  • Monosodium salt form suitable for aqueous and enzymatic studies.
  • Nominal purity 96.0% by HPLC; batch certificate of analysis may report higher purity.
  • White to off-white solid appearance; stable under recommended storage conditions.
  • Used in inflammation research, including hepcidin-ferroportin and IL-6/STAT3 pathway studies.
  • Molecular formula C10H14N5NaO11P2; molecular weight 465.18 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.