Learn More
Medchemexpress LLC Guanosine 5'-diphosphate sodium | 43139-22-6 | 99.9% | 465.18 | 500 MG
SDP
Catalog No. 5000436469
Encompass_Preferred
Guanosine 5'-diphosphate sodium is a nucleoside diphosphate known for activating adenosine 5'-triphosphate-sensitive K+ channels. It acts as an iron mobilizer by forming a complex with hepcidin, which inhibits the hepcidin-ferroportin (FPN) interaction and modulates the IL-6/Stat-3 pathway. This compound has been shown to ameliorate Turpentine-induced anemia of inflammation (AI) in mice, especially when combined with FeSO4, highlighting its utility in AI research.
  • Activates ATP-sensitive K+ channels.
  • Mobilizes iron by inhibiting hepcidin-FPN interaction.
  • Modulates IL-6/Stat-3 pathway.
  • Ameliorates anemia of inflammation (AI).
  • Increases iron absorption and FPN levels in liver, spleen, and enterocytes.
  • Reduces iron storage ferritin and Stat-3 phosphorylation.
  • Decreases Hamp mRNA and hepcidin expression.
Catalog No. 50-004-36469 Supplier Medchemexpress LLC Supplier No. HY113066C500MG
Guanosine 5'-diphosphate sodium is a nucleoside diphosphate known for activating adenosine 5'-triphosphate-sensitive K+ channels. It acts as an iron mobilizer by forming a complex with hepcidin, which inhibits the hepcidin-ferroportin (FPN) interaction and modulates the IL-6/Stat-3 pathway. This compound has been shown to ameliorate Turpentine-induced anemia of inflammation (AI) in mice, especially when combined with FeSO4, highlighting its utility in AI research.
  • Activates ATP-sensitive K+ channels.
  • Mobilizes iron by inhibiting hepcidin-FPN interaction.
  • Modulates IL-6/Stat-3 pathway.
  • Ameliorates anemia of inflammation (AI).
  • Increases iron absorption and FPN levels in liver, spleen, and enterocytes.
  • Reduces iron storage ferritin and Stat-3 phosphorylation.
  • Decreases Hamp mRNA and hepcidin expression.