Learn More
Medchemexpress LLC Guanosine 5'-diphospho-beta-L-fucose | 15839-70-0 | MFCD00058553 | 99.8% | 589.34 | C16H25N5O15P2 | 5 MG
SDP
Catalog No. 5000223070
Encompass_Preferred
GDP-L-fucose is a nucleotide sugar that functions as the activated donor of L-fucose in enzymatic fucosylation reactions. It is provided as a high-purity biochemical reagent for in vitro enzymology, glycoengineering, and metabolic pathway studies.
  • High purity suitable for biochemical assays.
  • Water-soluble nucleotide sugar for enzymatic reactions.
  • Serves as donor substrate for fucosyltransferases.
  • Useful for glycosylation and oligosaccharide biosynthesis studies.
  • Available in small pack sizes tailored for research use.
Catalog No. 50-002-23070 Supplier Medchemexpress LLC Supplier No. HY1344335MG
GDP-L-fucose is a nucleotide sugar that functions as the activated donor of L-fucose in enzymatic fucosylation reactions. It is provided as a high-purity biochemical reagent for in vitro enzymology, glycoengineering, and metabolic pathway studies.
  • High purity suitable for biochemical assays.
  • Water-soluble nucleotide sugar for enzymatic reactions.
  • Serves as donor substrate for fucosyltransferases.
  • Useful for glycosylation and oligosaccharide biosynthesis studies.
  • Available in small pack sizes tailored for research use.