Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanosine-5'-monophosphate, Free acid, ≥98%, MP Biomedicals™

Catalog No. ICN10190890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10190890 500 mg
ICN10190880 100 mg
ICN10190891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10190890 Supplier MP Biomedicals, Inc Supplier No. 0210190890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-32-5
Molecular Formula C10H14N5O8P
Molecular Weight (g/mol) 363.223
InChI Key RQFCJASXJCIDSX-UUOKFMHZSA-N
Synonym 5'-guanylic acid, guanosine monophosphate, guanylic acid, guanosine-5'-monophosphate, guanosine 5'-monophosphate, 5'-gmp, guanosine 5'-phosphate, guanylate, guanidine monophosphate, guanosine-phosphate
PubChem CID 6804
ChEBI CHEBI:17345
IUPAC Name [(2R,3S,4R,5R)-5-(2-amino-6-oxo-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl dihydrogen phosphate
SMILES C1=NC2=C(N1C3C(C(C(O3)COP(=O)(O)O)O)O)NC(=NC2=O)N

Specifications

Color White
Assay Percent Range ≥98%
Quantity 500 mg
Formula Weight 363.2
Physical Form Powder
Chemical Name or Material Guanosine-5'-monophosphate, FTIR
EINECSNumber 201-598-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.