Learn More
Abcam Guanosine 5'-monophosphate [GMP] disodium salt, ribonucleoside monophosphate, 25G
Catalog No. 502704587
Encompass_Preferred
MW 407.19 g/mol, Purity >98%. Ribonucleoside monophosphate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s). Can be used to study modulation of glutamatergic neurotransmission.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4587 Supplier Abcam Supplier No. AB14653425G
MW 407.19 g/mol, Purity >98%. Ribonucleoside monophosphate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s). Can be used to study modulation of glutamatergic neurotransmission.
The product is subject to the following: Abcam Restricted Use Statement