Learn More
Abcam Guanosine 5'-monophosphate [GMP] disodium salt, ribonucleoside monophosphate, 5G
Catalog No. 502704586
Encompass_Preferred
MW 407.19 g/mol, Purity >98%. Ribonucleoside monophosphate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s). Can be used to study modulation of glutamatergic neurotransmission.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4586 Supplier Abcam Supplier No. AB1465345G
MW 407.19 g/mol, Purity >98%. Ribonucleoside monophosphate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s). Can be used to study modulation of glutamatergic neurotransmission.
The product is subject to the following: Abcam Restricted Use Statement