Learn More
Medchemexpress LLC Guanosine 5'-(tetrahydrogen triphosphate), 2'-deoxy-2'-fluoro-2'-methyl-, sodium salt | 1621884-18-1 | MFCD20659124 | 99.7% | 627.13 g/mol | C11H13FN5Na4O13P3 | 5 MG
SDP
Catalog No. 5000221468
Encompass_Preferred
AT-9010 tetrasodium is the active triphosphate metabolite of AT-527 that acts as a nucleotide analog inhibitor of the NiRAN domain required for coronavirus RNA replication. It is used in biochemical and virology research to evaluate antiviral activity and mechanism of polymerase inhibition.
  • Active triphosphate metabolite that inhibits the NiRAN domain essential for viral replication.
  • Demonstrated inhibition of SARS-CoV-2 replication in research studies.
  • High analytical purity suitable for sensitive biochemical assays.
  • Appropriate for in vitro virology and enzymatic studies as a nucleotide analog.
  • Supplied in small research quantities for controlled experiments.
  • Requires storage at -80°C, protected from light, under inert gas for stability.
Catalog No. 50-002-21468 Supplier Medchemexpress LLC Supplier No. HY139165A5MG
AT-9010 tetrasodium is the active triphosphate metabolite of AT-527 that acts as a nucleotide analog inhibitor of the NiRAN domain required for coronavirus RNA replication. It is used in biochemical and virology research to evaluate antiviral activity and mechanism of polymerase inhibition.
  • Active triphosphate metabolite that inhibits the NiRAN domain essential for viral replication.
  • Demonstrated inhibition of SARS-CoV-2 replication in research studies.
  • High analytical purity suitable for sensitive biochemical assays.
  • Appropriate for in vitro virology and enzymatic studies as a nucleotide analog.
  • Supplied in small research quantities for controlled experiments.
  • Requires storage at -80°C, protected from light, under inert gas for stability.