Learn More
Medchemexpress LLC Guanosine 5'-(trihydrogen diphosphate), 2'-deoxy-, sodium salt (1:3) | 102783-74-4 | 98.8% | 493.15 | 25 MG
SDP
Catalog No. 5000366824
Encompass_Preferred
2′-Deoxy-GDP (dGDP) trisodium is an analog of guanosine monophosphate (GMP) that acts as an inhibitor of hypoxanthine phosphoribosyltransferase (HGPRT). This enzyme is crucial for the purine recycling pathway in all protozoan parasites. It has an IC50 of 12.5 μM.
  • Guanosine monophosphate (GMP) analogue.
  • Hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor.
  • IC50 of 12.5 μM for HGPRT.
  • Useful for research purposes.
Catalog No. 50-003-66824 Supplier Medchemexpress LLC Supplier No. 25MGHYW013111
2′-Deoxy-GDP (dGDP) trisodium is an analog of guanosine monophosphate (GMP) that acts as an inhibitor of hypoxanthine phosphoribosyltransferase (HGPRT). This enzyme is crucial for the purine recycling pathway in all protozoan parasites. It has an IC50 of 12.5 μM.
  • Guanosine monophosphate (GMP) analogue.
  • Hypoxanthine phosphoribosyltransferase (HGPRT) inhibitor.
  • IC50 of 12.5 μM for HGPRT.
  • Useful for research purposes.