Learn More
Medchemexpress LLC HY-W010737 250mg , Guanosine-5'-triphosphate (disodium salt) CAS:56001-37-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW010737250MG
Encompass_Preferred
HY-W010737 250mg Guanosine-5'-triphosphate (disodium salt) CAS: 56001-37-7 Guanosine-5'-triphosphate disodium salt (5'-GTP trisodium salt) is an activator of the signal transducing G proteins and also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1227