Learn More
Medchemexpress LLC Guanosine 5'-triphosphate-15N5 (dilithium) | 95.0% | 540.01 | C10H14Li215N5O14P3 | 5 MG
SDP
Catalog No. 5000263923
Encompass_Preferred
Guanosine 5'-triphosphate-15N5 (dilithium) is an isotopically labeled nucleotide supplied as the dilithium salt for research use. It is provided as a 5 mg aliquot (prepared at approx. 100 mM in Tris-HCl/H2O, pH 7.5) and is intended for tracer experiments, mass-spectrometry standards, and enzymatic or structural biology assays that require 15N-labeled GTP. Purity is listed as 95.0%.
  • Isotopically labeled (15N5) for tracer and mass spectrometry applications.
  • Dilithium salt form for improved stability and solubility.
  • Prepared at ~100 mM in Tris-HCl/H2O buffer, pH 7.5, ready for small-scale assays.
  • High purity (95.0%) suitable for analytical workflows.
  • Appropriate for enzymatic assays and structural biology studies.
  • Supplied as a small 5 mg quantity for research and analytical applications.
Catalog No. 50-002-63923 Supplier Medchemexpress LLC Supplier No. HY113225S35MG
Guanosine 5'-triphosphate-15N5 (dilithium) is an isotopically labeled nucleotide supplied as the dilithium salt for research use. It is provided as a 5 mg aliquot (prepared at approx. 100 mM in Tris-HCl/H2O, pH 7.5) and is intended for tracer experiments, mass-spectrometry standards, and enzymatic or structural biology assays that require 15N-labeled GTP. Purity is listed as 95.0%.
  • Isotopically labeled (15N5) for tracer and mass spectrometry applications.
  • Dilithium salt form for improved stability and solubility.
  • Prepared at ~100 mM in Tris-HCl/H2O buffer, pH 7.5, ready for small-scale assays.
  • High purity (95.0%) suitable for analytical workflows.
  • Appropriate for enzymatic assays and structural biology studies.
  • Supplied as a small 5 mg quantity for research and analytical applications.