Learn More
Medchemexpress LLC Guanosine 5'-triphosphate-5'-adenosine triammonium | 00-00-0 | 100.0% | 823.50 | C20H36N13O17P3 | 5 MG
SDP
Catalog No. 5000222534
Encompass_Preferred
Guanosine 5'-triphosphate-5'-adenosine triammonium is a 5′ cap analog and fluorescent substrate analog used in vitro for RNA synthesis and enzymatic assays. It is supplied as a high-purity triammonium salt in small-mass vials for research applications.
  • Cap analog for in vitro RNA synthesis.
  • Fluorescent substrate analog suitable for enzymatic assays.
  • High purity for reproducible experimental results.
  • Available in multiple small sizes, including 5 mg vials.
  • Molecular weight provided for accurate molarity calculations.
  • Triammonium salt form enhances aqueous solubility.
Catalog No. 50-002-22534 Supplier Medchemexpress LLC Supplier No. HY139101A5MG
Guanosine 5'-triphosphate-5'-adenosine triammonium is a 5′ cap analog and fluorescent substrate analog used in vitro for RNA synthesis and enzymatic assays. It is supplied as a high-purity triammonium salt in small-mass vials for research applications.
  • Cap analog for in vitro RNA synthesis.
  • Fluorescent substrate analog suitable for enzymatic assays.
  • High purity for reproducible experimental results.
  • Available in multiple small sizes, including 5 mg vials.
  • Molecular weight provided for accurate molarity calculations.
  • Triammonium salt form enhances aqueous solubility.