Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine 5'-triphosphate-5'-adenosine triammonium | 00-00-0 | 100.0% | C20H36N13O17P3 | 10MG
SDP

Catalog No. 5000260726
Encompass_Preferred

Guanosine 5'-triphosphate-5'-adenosine triammonium is a 5′-cap analog and fluorescent substrate analog used as a cap structure mimic in in vitro RNA synthesis and related biochemical assays. It is supplied as the triammonium salt and accompanied by analytical documentation for quality verification.

  • High purity (99.97%).
  • Molecular weight 823.50.
  • Chemical formula C20H36N13O17P3.
  • Available in 1 mg, 5 mg, and 10 mg sizes (larger sizes by inquiry).
  • Includes data sheet, SDS, COA, HNMR, CNMR, RP-HPLC, and MS documents.
  • Suitable as a cap analog for in vitro transcription and biochemical assays.

Catalog No. 50-002-60726 Supplier Medchemexpress LLC Supplier No. HY139101A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanosine 5'-triphosphate-5'-adenosine triammonium is a 5′-cap analog and fluorescent substrate analog used as a cap structure mimic in in vitro RNA synthesis and related biochemical assays. It is supplied as the triammonium salt and accompanied by analytical documentation for quality verification.

  • High purity (99.97%).
  • Molecular weight 823.50.
  • Chemical formula C20H36N13O17P3.
  • Available in 1 mg, 5 mg, and 10 mg sizes (larger sizes by inquiry).
  • Includes data sheet, SDS, COA, HNMR, CNMR, RP-HPLC, and MS documents.
  • Suitable as a cap analog for in vitro transcription and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.