Learn More
Medchemexpress LLC Guanosine 5'-triphosphate-5'-adenosine triammonium | 99.97% | 823.50 | 25 MG
SDP
Catalog No. 5000348102
Encompass_Preferred
Guanosine 5'-triphosphate-5'-adenosine (GpppA) triammonium is a 5' cap analog that can be used for RNA synthesis in vitro. It is also described as a fluorescent substrate analog.
  • Fluorescent substrate analog.
  • Can be used for RNA synthesis in vitro.
  • Acts as an artificial substrate when labeled with pyrene at the 3′-O position of adenosine.
  • Fluorescently labeled GpppA triammonium and GpppG analogs are potential substrates that balance structural complexity and enzymatic requirements.
Catalog No. 50-003-48102 Supplier Medchemexpress LLC Supplier No. 25MGHY139101A
Guanosine 5'-triphosphate-5'-adenosine (GpppA) triammonium is a 5' cap analog that can be used for RNA synthesis in vitro. It is also described as a fluorescent substrate analog.
  • Fluorescent substrate analog.
  • Can be used for RNA synthesis in vitro.
  • Acts as an artificial substrate when labeled with pyrene at the 3′-O position of adenosine.
  • Fluorescently labeled GpppA triammonium and GpppG analogs are potential substrates that balance structural complexity and enzymatic requirements.