Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Guanosine-5'-triphosphate Disodium Salt, ≥95%, MP Biomedicals™
Shop All MP Biomedicals, Inc ProductsDescription
- Guanosine 5′-triphosphate (GTP), a purine trinucleotide, is a substrate for RNA polymerases and a wide range of GTPase including G-protein coupled receptor (GPCR) GTPases and cell signaling and cycling associated guanine nucleotide exchange factors (GEF).
Specifications
Specifications
| Absorbance | λ max (water): 252 ± 3 nm |
| Color | White |
| Assay Percent Range | ≥95% |
| Quantity | 500 mg |
| IUPAC Name | ({[({[(2R,3S,4R,5R)-5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid magnesium |
| Formula Weight | 567.2 |
| Percent Purity | ≥95% |
| Physical Form | Powder |
| Chemical Name or Material | Guanosine-5'-triphosphate disodium salt, For HPLC analysis |
Safety and Handling
| EINECSNumber | 259-940-7 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?