Learn More
Sigma Aldrich Fine Chemicals Biosciences Guanosine 5′-triphosphate sodium salt hydrate, 36051-31-7, MFCD00077781, 250 mg
Catalog No. NC0623123
Encompass
Empirical Formula (Hill Notation): C10H16N5O14P3 · xNa+, Molecular Weight: 523.18 (free acid basis), ≥95% (HPLC), powder, Synonym: GTP. Guanosine 5′-triphosphate sodium salt hydrate has been used: in the preparation of horseradish peroxidase (HRP)-modified platination reagents, in whole cell, voltage-clamp recordings, and in in vitro inhibition of transglutaminase (tTG).
Catalog No. NC0623123 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G8877250MG
Empirical Formula (Hill Notation): C10H16N5O14P3 · xNa+, Molecular Weight: 523.18 (free acid basis), ≥95% (HPLC), powder, Synonym: GTP. Guanosine 5′-triphosphate sodium salt hydrate has been used: in the preparation of horseradish peroxidase (HRP)-modified platination reagents, in whole cell, voltage-clamp recordings, and in in vitro inhibition of transglutaminase (tTG).