Learn More
Guanosine 5'-Triphosphate, Sodium Salt, Spectrum™ Chemical
Shop All Spectrum Chemical Mfg. Corp. Products

Description
GTP Sodium Salt, Guanosine 5'-Triphosphate, Periodate Oxidized Sodium Salt
Specifications
Specifications
| Packaging | Bottle |
| Quantity | 1 g |
| Molecular Weight (g/mol) | 545.16 |
| Grade | Ungraded |
| Physical Form | Crystalline Powder or Lumps |
| Chemical Name or Material | Guanosine 5'-Triphosphate, Sodium Salt |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.