Learn More
TARGETMOL CHEMICALS INC Guanosine 5-triphosphate triso
SDP
Catalog No. 502878666
Encompass_Preferred
Also available in 1 g, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Guanosine 5'-triphosphate trisodium salt (5'-GTP trisodium salt) activates signal-transducing G proteins involved in cellular processes such as proliferation, differentiation, and activation of intracellular kinase cascades. It regulates proliferation and apoptosis through the hydrolysis by small GTPases Ras and Rho, and plays a role in vesicle docking, fusion, and formation via Rab. Additionally, GTP serves as an energy-rich precursor for the biosynthesis of DNA and RNA. Purity 99.45%
Catalog No. 50-287-8666 Supplier TARGETMOL CHEMICALS INC Supplier No. T4969500MG
Also available in 1 g, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Guanosine 5'-triphosphate trisodium salt (5'-GTP trisodium salt) activates signal-transducing G proteins involved in cellular processes such as proliferation, differentiation, and activation of intracellular kinase cascades. It regulates proliferation and apoptosis through the hydrolysis by small GTPases Ras and Rho, and plays a role in vesicle docking, fusion, and formation via Rab. Additionally, GTP serves as an energy-rich precursor for the biosynthesis of DNA and RNA. Purity 99.45%