Learn More
Medchemexpress LLC Guanosine 5'-triphosphate (trisodium salt) | 36051-31-7 | 95.0% | 1 G
SDP
Catalog No. 5000341518
Encompass_Preferred
Guanosine 5'-triphosphate (5'-GTP) trisodium salt is a G protein signaling activator and a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA. It promotes myogenic cell differentiation by upregulating miRNA (miR133a, miR133b) and myogenic regulatory factor expression, and by inducing human myogenic precursor cells to release exosomes containing guanosine molecules. This product holds promise for research in biosynthesis and skeletal muscle regeneration.
  • G protein signaling activator
  • High-energy precursor in biosynthesis of nucleotide units in DNA and RNA
  • Promotes myogenic cell differentiation
  • Induces release of exosomes from human myogenic precursor cells
  • Suitable for research in biosynthesis
  • Suitable for research in skeletal muscle regeneration
Catalog No. 50-003-41518 Supplier Medchemexpress LLC Supplier No. HY12695B1G
Guanosine 5'-triphosphate (5'-GTP) trisodium salt is a G protein signaling activator and a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA. It promotes myogenic cell differentiation by upregulating miRNA (miR133a, miR133b) and myogenic regulatory factor expression, and by inducing human myogenic precursor cells to release exosomes containing guanosine molecules. This product holds promise for research in biosynthesis and skeletal muscle regeneration.
  • G protein signaling activator
  • High-energy precursor in biosynthesis of nucleotide units in DNA and RNA
  • Promotes myogenic cell differentiation
  • Induces release of exosomes from human myogenic precursor cells
  • Suitable for research in biosynthesis
  • Suitable for research in skeletal muscle regeneration