Learn More
Medchemexpress LLC Guanosine 5'-triphosphate (trisodium salt) | 36051-31-7 | 95.0% | 1 G
SDP
Catalog No. 5000341559
Encompass_Preferred
Guanosine 5'-triphosphate (5'-GTP) trisodium salt is a G protein signaling activator and a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA. It can promote myogenic cell differentiation by upregulating miRNA and myogenic regulatory factor expression, and by inducing human myogenic precursor cells to release exosomes containing guanosine molecules. It holds promise for research in biosynthesis and skeletal muscle regeneration.
  • Is a G protein signaling activator
  • Is a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA
  • Can promote myogenic cell differentiation
  • Upregulates miRNA (miR133a, miR133b) and myogenic regulatory factor expression
  • Induces human myogenic precursor cells to release exosomes containing guanosine molecules
  • Holds promise for research in biosynthesis and skeletal muscle regeneration
  • For research use only
Catalog No. 50-003-41559 Supplier Medchemexpress LLC Supplier No. HY126951G
Guanosine 5'-triphosphate (5'-GTP) trisodium salt is a G protein signaling activator and a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA. It can promote myogenic cell differentiation by upregulating miRNA and myogenic regulatory factor expression, and by inducing human myogenic precursor cells to release exosomes containing guanosine molecules. It holds promise for research in biosynthesis and skeletal muscle regeneration.
  • Is a G protein signaling activator
  • Is a high-energy precursor in the biosynthesis of nucleotide units in DNA and RNA
  • Can promote myogenic cell differentiation
  • Upregulates miRNA (miR133a, miR133b) and myogenic regulatory factor expression
  • Induces human myogenic precursor cells to release exosomes containing guanosine molecules
  • Holds promise for research in biosynthesis and skeletal muscle regeneration
  • For research use only