Learn More
Medchemexpress LLC Guanosine 5'-triphosphate trisodium salt hydrate | 207300-85-4 | 95.0% | 100 MG
SDP
Catalog No. 5000341538
Encompass_Preferred
Guanosine 5'-triphosphate trisodium salt hydrate is an activator of the signal transducing G proteins and serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.
  • Activator of signal transducing G proteins
  • Energy-rich precursor in DNA and RNA biosynthesis
  • For research use only
Catalog No. 50-003-41538 Supplier Medchemexpress LLC Supplier No. HY12695B100MG
Guanosine 5'-triphosphate trisodium salt hydrate is an activator of the signal transducing G proteins and serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA.
  • Activator of signal transducing G proteins
  • Energy-rich precursor in DNA and RNA biosynthesis
  • For research use only