Learn More
Medchemexpress LLC Guanosine 5'-triphosphate trisodium salt hydrate | 207300-85-4 | 95.0% | 200 MG
SDP
Catalog No. 5000342175
Encompass_Preferred
Guanosine 5'-triphosphate trisodium salt hydrate is an activator of the signal transducing G proteins. It also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA. The chemical formula is C10H13N5Na3O14P3·xH2O and it appears as a white to off-white solid.
  • Activator of signal transducing G proteins
  • Energy-rich precursor of mononucleotide units
  • Involved in enzymatic biosynthesis of DNA and RNA
  • Appears as a white to off-white solid
Catalog No. 50-003-42175 Supplier Medchemexpress LLC Supplier No. HY12695B200MG
Guanosine 5'-triphosphate trisodium salt hydrate is an activator of the signal transducing G proteins. It also serves as an energy-rich precursor of mononucleotide units in the enzymatic biosynthesis of DNA and RNA. The chemical formula is C10H13N5Na3O14P3·xH2O and it appears as a white to off-white solid.
  • Activator of signal transducing G proteins
  • Energy-rich precursor of mononucleotide units
  • Involved in enzymatic biosynthesis of DNA and RNA
  • Appears as a white to off-white solid