Learn More
Medchemexpress LLC Guanosine 5'-triphosphate trisodium salt hydrate | 207300-85-4 | 95.0% | 50 MG
SDP
Catalog No. 5000341573
Encompass_Preferred
Guanosine 5'-triphosphate trisodium salt hydrate (5'-GTP trisodium salt hydrate) is a chemical reagent that functions as an activator for signal transducing G proteins. It also serves as an energy-rich precursor of mononucleotide units, which are essential for the enzymatic biosynthesis of DNA and RNA. This product is intended for research use only.
  • Functions as an activator for signal transducing G proteins.
  • Serves as an energy-rich precursor of mononucleotide units.
  • Essential for the enzymatic biosynthesis of DNA and RNA.
  • Intended for research use only.
Catalog No. 50-003-41573 Supplier Medchemexpress LLC Supplier No. HY12695B50MG
Guanosine 5'-triphosphate trisodium salt hydrate (5'-GTP trisodium salt hydrate) is a chemical reagent that functions as an activator for signal transducing G proteins. It also serves as an energy-rich precursor of mononucleotide units, which are essential for the enzymatic biosynthesis of DNA and RNA. This product is intended for research use only.
  • Functions as an activator for signal transducing G proteins.
  • Serves as an energy-rich precursor of mononucleotide units.
  • Essential for the enzymatic biosynthesis of DNA and RNA.
  • Intended for research use only.