Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine 5'-triphosphate trisodium salt hydrate | 207300-85-4 | 95.0% | 500 MG
SDP

Catalog No. 5000340826
Encompass_Preferred

Guanosine 5'-triphosphate trisodium salt hydrate activates signal transducing G proteins and functions as an energy-rich precursor for mononucleotide units in the enzymatic biosynthesis of DNA and RNA. This product is intended for research use only.

  • Activates signal transducing G proteins
  • Precursor for DNA and RNA biosynthesis
  • Solid, white to off-white physical form
  • Store at -20°C under nitrogen, away from moisture
  • When in solvent, stable at -80°C for 6 months and -20°C for 1 month
  • Soluble in H2O at ≥ 150 mg/mL

Catalog No. 50-003-40826 Supplier Medchemexpress LLC Supplier No. HY12695B500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanosine 5'-triphosphate trisodium salt hydrate activates signal transducing G proteins and functions as an energy-rich precursor for mononucleotide units in the enzymatic biosynthesis of DNA and RNA. This product is intended for research use only.

  • Activates signal transducing G proteins
  • Precursor for DNA and RNA biosynthesis
  • Solid, white to off-white physical form
  • Store at -20°C under nitrogen, away from moisture
  • When in solvent, stable at -80°C for 6 months and -20°C for 1 month
  • Soluble in H2O at ≥ 150 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.