Learn More
Apexbio Technology LLC GUANOSINE-5-TRIPO4 DINA-500mg
SDP
Catalog No. 5000897325
Encompass_Preferred
Guanosine-5 -triphosphate disodium salt (CAS No 56001-37-7) is a nucleotide commonly used in biochemical and molecular biology research Originating from cellular processes it acts as an energy-rich molecule involved in metabolic reactions including protein synthesis and cell signaling pathways such as G-protein-coupled receptor (GPCR) activation Its activity is typically observed in the nanomolar to low micromolar range making it suitable for various applications This compound is frequently used in vitro to study signal transduction enzyme kinetics and cellular metabolism and is applied in cell models and animal studies to evaluate biochemical pathways and drug screening processes Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97325 Supplier Apexbio Technology LLC Supplier No. C8653500
Guanosine-5 -triphosphate disodium salt (CAS No 56001-37-7) is a nucleotide commonly used in biochemical and molecular biology research Originating from cellular processes it acts as an energy-rich molecule involved in metabolic reactions including protein synthesis and cell signaling pathways such as G-protein-coupled receptor (GPCR) activation Its activity is typically observed in the nanomolar to low micromolar range making it suitable for various applications This compound is frequently used in vitro to study signal transduction enzyme kinetics and cellular metabolism and is applied in cell models and animal studies to evaluate biochemical pathways and drug screening processes Experimental concentrations typically depend on the specific research design