Learn More
Medchemexpress LLC Guanosine, 6-O-methyl- | 7803-88-5 | 99.7% | 297.27 | 100 MG
SDP
Catalog No. 5000438723
Encompass_Preferred
6-O-Methyl Guanosine is a modified nucleoside that inhibits colony-forming ability in a malignant xeroderma pigmentosum cell line.
  • Modified nucleoside
  • Inhibits colony-forming ability in a malignant xeroderma pigmentosum cell line
  • For research use only
Catalog No. 50-004-38723 Supplier Medchemexpress LLC Supplier No. HY111648100MG
6-O-Methyl Guanosine is a modified nucleoside that inhibits colony-forming ability in a malignant xeroderma pigmentosum cell line.
  • Modified nucleoside
  • Inhibits colony-forming ability in a malignant xeroderma pigmentosum cell line
  • For research use only