Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine, 7-methyl-, iodide (1:1) | 81100-62-1 | 99.6% | 100MG
SDP

Catalog No. 5000234734
Encompass_Preferred

Guanosine, 7-methyl-, iodide (1:1) | 81100-62-1 | 99.6% | 100MG

Catalog No. 50-002-34734 Supplier Medchemexpress LLC Supplier No. HY122524A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guanosine, 7-methyl-, iodide (1:1) | 81100-62-1 | 99.6% | 100MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.