Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine, 8-bromo-2'-deoxy- | 13389-03-2 | ≥98.0% | 346.14 | 1 ML
SDP

Catalog No. 5000452816
Encompass_Preferred

8-Bromo-2'-deoxyguanosine is an inflammation-related DNA halogenated adduct and early biomarker of oxidative tissue damage. It serves as an immunogen for mAb8B3, detecting DNA modifications. Elevated urinary levels occur in inflammatory disease models. Used in studies for inflammatory diseases, diabetes, hepatocellular carcinoma, and UV-B induced skin inflammation.

  • Inflammation-related DNA halogenated adduct.
  • Early biomarker of inflammation-induced oxidative tissue damage.
  • Immunogen for mAb8B3 monoclonal antibody.
  • Urinary levels increase in inflammatory disease models.
  • Applications in inflammatory diseases, diabetes, hepatocellular carcinoma, UV-B induced skin inflammation.
  • High, specific cross-reactivity with mAb8B3 in competitive indirect ELISA.

Catalog No. 50-004-52816 Supplier Medchemexpress LLC Supplier No. HYW01116810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromo-2'-deoxyguanosine is an inflammation-related DNA halogenated adduct and early biomarker of oxidative tissue damage. It serves as an immunogen for mAb8B3, detecting DNA modifications. Elevated urinary levels occur in inflammatory disease models. Used in studies for inflammatory diseases, diabetes, hepatocellular carcinoma, and UV-B induced skin inflammation.

  • Inflammation-related DNA halogenated adduct.
  • Early biomarker of inflammation-induced oxidative tissue damage.
  • Immunogen for mAb8B3 monoclonal antibody.
  • Urinary levels increase in inflammatory disease models.
  • Applications in inflammatory diseases, diabetes, hepatocellular carcinoma, UV-B induced skin inflammation.
  • High, specific cross-reactivity with mAb8B3 in competitive indirect ELISA.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.