Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine, N-(1-oxobutyl)-, cyclic 3',5'-(hydrogen phosphate) 2'-butanoate, sodium salt (1:1) | 51116-00-8 | 99.7% | 50 MG
SDP

Catalog No. 5000338344
Encompass_Preferred

Dibutyryl-cGMP sodium (Bt2cGMP sodium) is a cell-permeable cGMP analogue that preferentially activates cGMP-dependent protein kinase (PKG). It inhibits the release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets and induces peripheral antinociception via activation of ATP-sensitive K+ channels. It is for research use only and not sold to patients.

  • Appearance: solid
  • Color: white to light yellow
  • Molecular weight: 507.37
  • Formula: C18H23N5NaO9P
  • Solubility in H2O: 100 mg/mL (197.09 mM; requires ultrasonic)
  • Solubility in DMSO: 100 mg/mL (197.09 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility)

Catalog No. 50-003-38344 Supplier Medchemexpress LLC Supplier No. HY13035450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dibutyryl-cGMP sodium (Bt2cGMP sodium) is a cell-permeable cGMP analogue that preferentially activates cGMP-dependent protein kinase (PKG). It inhibits the release of [3H]-arachidonic acid from γ thrombin-stimulated human platelets and induces peripheral antinociception via activation of ATP-sensitive K+ channels. It is for research use only and not sold to patients.

  • Appearance: solid
  • Color: white to light yellow
  • Molecular weight: 507.37
  • Formula: C18H23N5NaO9P
  • Solubility in H2O: 100 mg/mL (197.09 mM; requires ultrasonic)
  • Solubility in DMSO: 100 mg/mL (197.09 mM; requires ultrasonic; hygroscopic DMSO has a significant impact on solubility)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.