Learn More
Enzo Life Sciences Guanosine 5'-O-(3-thiotriphosphate) . tetralithium salt, (1mg), CAS Number: 94825-44-2
SDP
Supplier:  Enzo Life Sciences ALX480056M001
Encompass_Preferred
Nonhydrolyzable GTP analog. A nonhydrolyzable analog of guanosine 5’-O-triphosphate (GTP). Stimulates inositol trisphosphate formation in membranes. Alternative Name: GTP-gamma-S . 4Li. Formula: C10H12N5O13P3 S. 4Li. MW: 535.2 . 27.8. CAS Number: 94825-44-2. Purity: ≥85% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C
Catalog No. NC1101601