Learn More
Medchemexpress LLC Guanosine triphosphate-13C10 dilithium | 86-01-1 | 99.4% | 544.97 | 1 MG
SDP
Catalog No. 5000437249
Encompass_Preferred
Guanosine triphosphate-13C10 (GTP-13C10) dilithium is a 13C-labeled version of Guanosine triphosphate (HY-113225), which is a native nucleotide. Derivatives of GTP may act as specific inhibitors against COVID-19.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-37249 Supplier Medchemexpress LLC Supplier No. HY113225S51MG
Guanosine triphosphate-13C10 (GTP-13C10) dilithium is a 13C-labeled version of Guanosine triphosphate (HY-113225), which is a native nucleotide. Derivatives of GTP may act as specific inhibitors against COVID-19.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.