Learn More
Medchemexpress LLC Guanosine triphosphate-15N5,d14 (dilithium) | 86-01-1 | 98.4% | 554.10 | 1 MG
SDP
Catalog No. 5000436888
Encompass_Preferred
Guanosine triphosphate-15N5,d14 dilithium is a deuterium and 15N labeled form of Guanosine triphosphate, a native nucleotide. Derivatives of GTP may be used as specific inhibitors against COVID-19. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-36888 Supplier Medchemexpress LLC Supplier No. HY113225S41MG
Guanosine triphosphate-15N5,d14 dilithium is a deuterium and 15N labeled form of Guanosine triphosphate, a native nucleotide. Derivatives of GTP may be used as specific inhibitors against COVID-19. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS