Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanosine triphosphate-15N5, dilithium | 95.0% | 540.01 | 1 MG
SDP

Catalog No. 5000436116
Encompass_Preferred

Guanosine triphosphate-15N5 (GTP-15N5) dilithium is a 15N labeled version of Guanosine triphosphate, which is a native nucleotide. Derivatives of GTP can serve as specific inhibitors against COVID-19.

  • Labeled with 15N.
  • Purity of 95.0%.
  • Molecular weight is 540.01.
  • Chemical formula is C10H14Li215N5O14P3.
  • Appears as a liquid.
  • Color is colorless to light yellow.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles.

Catalog No. 50-004-36116 Supplier Medchemexpress LLC Supplier No. HY113225S31MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Guanosine triphosphate-15N5 (GTP-15N5) dilithium is a 15N labeled version of Guanosine triphosphate, which is a native nucleotide. Derivatives of GTP can serve as specific inhibitors against COVID-19.

  • Labeled with 15N.
  • Purity of 95.0%.
  • Molecular weight is 540.01.
  • Chemical formula is C10H14Li215N5O14P3.
  • Appears as a liquid.
  • Color is colorless to light yellow.
  • For research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration may affect pharmacokinetic and metabolic profiles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.