Learn More
Medchemexpress LLC Guanosine triphosphate trisodium salt | 36051-31-7 | 589.13 | 1 ML
SDP
Catalog No. 5000435682
Encompass_Preferred
Guanosine triphosphate trisodium salt, 100 mM Solution, PCR Grade is a solution prepared from guanosine triphosphate at a concentration of 100 mM. It is free of DNase, RNase, and phosphatase contamination, making it suitable for various molecular biology research applications.
  • Prepared from guanosine triphosphate at 100 mM concentration.
  • Free of DNase contamination.
  • Free of RNase contamination.
  • Free of phosphatase contamination.
  • Suitable for molecular biology research.
Catalog No. 50-004-35682 Supplier Medchemexpress LLC Supplier No. HY113225H1ML
Guanosine triphosphate trisodium salt, 100 mM Solution, PCR Grade is a solution prepared from guanosine triphosphate at a concentration of 100 mM. It is free of DNase, RNase, and phosphatase contamination, making it suitable for various molecular biology research applications.
  • Prepared from guanosine triphosphate at 100 mM concentration.
  • Free of DNase contamination.
  • Free of RNase contamination.
  • Free of phosphatase contamination.
  • Suitable for molecular biology research.