Learn More
Medchemexpress LLC Guanoxabenz hydrochloride | 23256-40-8 | 283.54 | 1 ML
SDP
Catalog No. 5000448437
Encompass_Preferred
Guanoxabenz hydrochloride is an α2 adrenergic receptor agonist supplied as a 10 mM solution in 1 mL of DMSO, primarily used for research purposes.
  • Functions as an α2 adrenergic receptor agonist.
  • Suitable for in vitro and in vivo research.
  • Provided as a 10 mM solution in DMSO.
  • Purity greater than 99.7%.
  • High-affinity binding inhibited by N-hydroxyguanidine analogs and metabolic inhibitors.
  • Known as a centrally active antihypertensive drug.
  • Causes dose-related reduction in locomotor activity.
Catalog No. 50-004-48437 Supplier Medchemexpress LLC Supplier No. HYU00123A10MM/1ML
Guanoxabenz hydrochloride is an α2 adrenergic receptor agonist supplied as a 10 mM solution in 1 mL of DMSO, primarily used for research purposes.
  • Functions as an α2 adrenergic receptor agonist.
  • Suitable for in vitro and in vivo research.
  • Provided as a 10 mM solution in DMSO.
  • Purity greater than 99.7%.
  • High-affinity binding inhibited by N-hydroxyguanidine analogs and metabolic inhibitors.
  • Known as a centrally active antihypertensive drug.
  • Causes dose-related reduction in locomotor activity.