Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanylate-binding protein 1, human (HEK293, His) | >95.0% | 50 UG
SDP

Catalog No. 5000293158
Encompass_Preferred

Recombinant human guanylate-binding protein 1 expressed in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution at high purity for research applications including enzymatic assays, binding studies, and antibody validation.

  • Expressed in HEK293 mammalian cells.
  • C-terminal 10xHis tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight ~65 kDa.
  • Supplied in 0.22 μm filtered PBS, pH 7.4.
  • Recommended storage at -80°C; avoid repeated freeze-thaw cycles.
  • Available in small research sizes including 50 μg.

Catalog No. 50-002-93158 Supplier Medchemexpress LLC Supplier No. HYP7517250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human guanylate-binding protein 1 expressed in HEK293 cells with a C-terminal His tag. Supplied as a 0.22 μm filtered PBS solution at high purity for research applications including enzymatic assays, binding studies, and antibody validation.

  • Expressed in HEK293 mammalian cells.
  • C-terminal 10xHis tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight ~65 kDa.
  • Supplied in 0.22 μm filtered PBS, pH 7.4.
  • Recommended storage at -80°C; avoid repeated freeze-thaw cycles.
  • Available in small research sizes including 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.