Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Guanylate-binding protein 1 | >95.0% | 2 UG
SDP

Catalog No. 5000291749
Encompass_Preferred

Recombinant human guanylate-binding protein 1 (GBP1) expressed in HEK293 cells and supplied as a C-terminal His-tagged protein in PBS. GBP1 is a large GTPase involved in innate immunity, inflammasome regulation, and antiviral responses; this high-purity preparation is suitable for biochemical and cell-based assays.

  • Expressed in HEK293 cells
  • C-terminal 10xHis tag for detection and purification
  • Purity >95% by reducing SDS-PAGE
  • Approximate molecular weight 65 kDa
  • Supplied in 0.22 μm filtered PBS, pH 7.4
  • Shipped on dry ice; recommended storage at -80°C
  • Available in small packs down to 2 μg for assay optimization

Catalog No. 50-002-91749 Supplier Medchemexpress LLC Supplier No. HYP751722UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human guanylate-binding protein 1 (GBP1) expressed in HEK293 cells and supplied as a C-terminal His-tagged protein in PBS. GBP1 is a large GTPase involved in innate immunity, inflammasome regulation, and antiviral responses; this high-purity preparation is suitable for biochemical and cell-based assays.

  • Expressed in HEK293 cells
  • C-terminal 10xHis tag for detection and purification
  • Purity >95% by reducing SDS-PAGE
  • Approximate molecular weight 65 kDa
  • Supplied in 0.22 μm filtered PBS, pH 7.4
  • Shipped on dry ice; recommended storage at -80°C
  • Available in small packs down to 2 μg for assay optimization

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.